Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020616.1 | NP_065641.1 | 1 - 2 | 39 | 58 | |
GenBank mRNA | AK086219.1 | - | 1 - 2 | 72 | 58 | |
AK155010.1 | - | 1 - 2 | 87 | 58 | ||
AK159903.1 | - | 1 - 2 | 40 | 58 | ||
AK163012.1 | - | 1 - 2 | 86 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |