Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008080.5 | NP_032106.1 | 9 - 10 | 1508 | 71 | |
NM_027739.2 | NP_082015.2 | 8 - 9 | 1317 | 71 | ||
NR_045166.1 | 8 - 9 | 1327 | 71 | |||
GenBank mRNA | AK144484.1 | - | 9 - 10 | 1474 | 71 | |
BC019969.1 | - | 8 - 9 | 1258 | 71 | ||
BC036996.1 | - | 8 - 9 | 1242 | 5038560 | 71 | |
BC057199.1 | - | 9 - 10 | 1439 | 5315443 | 71 | |
L25885.1 | - | 8 - 9 | 1271 | 71 | ||
U18975.1 | - | 8 - 9 | 1146 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |