Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001295.2 | NP_001001295.2 | 11 - 12 | 1793 | 126 | |
NM_001177784.1 | NP_001171255.1 | 11 - 12 | 1754 | 126 | ||
NM_172519.3 | NP_766107.1 | 11 - 12 | 1697 | 126 | ||
GenBank mRNA | AK029974.1 | - | 11 - 12 | 1698 | 126 | |
AK032451.1 | - | 11 - 12 | 1702 | 126 | ||
AK077031.1 | - | 11 - 12 | 1757 | 126 | ||
AK150851.1 | - | 11 - 12 | 1743 | 126 | ||
AK151780.1 | - | 11 - 12 | 1743 | 126 | ||
AK153433.1 | - | 11 - 12 | 1743 | 126 | ||
AK173310.1 | - | 11 - 12 | 1727 | 126 | ||
BC056939.1 | - | 11 - 12 | 1726 | 5708685 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |