Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010124.2 | NP_034254.1 | 1 - 2 | 266 | 104 | |
GenBank mRNA | AK133912.1 | - | 1 - 2 | 154 | 104 | |
AK148374.1 | - | 305 | 104 | |||
BC015082.1 | - | 2 - 3 | 247 | 4218820 | 104 | |
BC119539.2 | - | 2 - 3 | 230 | 40056467 | 104 | |
U75530.1 | - | 1 - 2 | 263 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |