Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027823.1 | NP_082099.1 | 1 - 2 | 394 | 77 | |
GenBank mRNA | AK018520.1 | - | 1 - 2 | 408 | 77 | |
AK082857.1 | - | 1 - 2 | 483 | 77 | ||
BC137891.1 | - | 1 - 2 | 394 | 8860910 | 77 | |
BC137892.1 | - | 1 - 2 | 394 | 8860911 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |