Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2158841 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
512616 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842984 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611356 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
419849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359109 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
189786 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776999 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
722697 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802555 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
961123 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93220 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
192447 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416643 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
110041 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
827502 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
597275 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576695 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
AK084047.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
AK087479.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
AK149202.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
BC132529.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
NM_001024147.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319545 |
NM_173403.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399817 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391966 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
578249 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
35549 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501421 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
54303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
13514 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
37437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
90034 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
166517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
164096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
85936 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P112626 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P167461 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P103037 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2158841 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07216877 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112626 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P167461 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P103037 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P292566 |
AK003432.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323369 |
AK029113.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323369 |
BC094337.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323369 |
NM_001081056.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323369 |
AK014235.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131828 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P103037 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P167461 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112626 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08377787 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08377788 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08377795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27504556 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08565813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504554 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504511 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504607 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504576 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504352 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504515 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08500740 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504520 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504569 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504572 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08967963 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08484824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504517 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504509 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504559 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504579 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504598 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504380 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504573 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08719177 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504564 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504594 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504506 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504568 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504379 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504610 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08313265 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504604 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08594035 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504373 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504581 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504590 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08548687 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08902614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504621 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504513 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504561 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504570 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504556 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504558 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504612 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504377 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504383 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504585 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08484825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08659684 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504372 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504523 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504552 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504593 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27504516 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08620920 |
- |
- |