Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030596.3 | NP_085099.2 | 6 - 7 | 751 | 62 | |
GenBank mRNA | AK029018.1 | - | 773 | 62 | ||
AK037142.1 | - | 6 - 7 | 782 | 62 | ||
AK148657.1 | - | 6 - 7 | 752 | 62 | ||
BC132058.1 | - | 6 - 7 | 780 | 40130335 | 62 | |
BC132639.1 | - | 6 - 7 | 780 | 40130916 | 62 | |
U86016.1 | - | 6 - 7 | 680 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |