Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011397.4 | NP_035527.3 | 12 - 13 | 1562 | 66 | |
GenBank mRNA | AF058318.1 | - | 12 - 13 | 1472 | 66 | |
AK085499.1 | - | 12 - 13 | 1477 | 66 | ||
AK143959.1 | - | 12 - 13 | 1563 | 66 | ||
AK165439.1 | - | 12 - 13 | 1498 | 66 | ||
BC013528.1 | - | 12 - 13 | 1491 | 4238222 | 66 | |
BE990394.1 | - | 1 - 2 | 160 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |