Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012042.3 | NP_036172.1 | 11 - 12 | 70 | ||
GenBank mRNA | AF136441.1 | - | 10 - 11 | 1476 | 70 | |
AF176910.1 | - | 11 - 12 | 70 | |||
AK147004.1 | - | 11 - 12 | 70 | |||
AK164274.1 | - | 11 - 12 | 70 | |||
AK165978.1 | - | 11 - 12 | 70 | |||
BC004836.1 | - | 10 - 11 | 1430 | 70 | ||
BC029260.1 | - | 11 - 12 | 2812400 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |