Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012042.3 | NP_036172.1 | 12 - 13 | 1671 | 105 | |
GenBank mRNA | AF136441.1 | - | 11 - 12 | 1526 | 105 | |
AF176910.1 | - | 12 - 13 | 1560 | 105 | ||
AK147004.1 | - | 12 - 13 | 1653 | 105 | ||
AK164274.1 | - | 12 - 13 | 1638 | 105 | ||
AK165978.1 | - | 12 - 13 | 1612 | 105 | ||
BC004836.1 | - | 11 - 12 | 1480 | 105 | ||
BC029260.1 | - | 12 - 13 | 1671 | 2812400 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |