Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028679.3 | NP_082955.2 | 7 - 8 | 885 | 63 | |
GenBank mRNA | AF461763.1 | - | 7 - 8 | 865 | 63 | |
AJ440757.2 | - | 7 - 8 | 807 | 63 | ||
AK014783.1 | - | 7 - 8 | 936 | 63 | ||
AK028436.1 | - | 798 | 63 | |||
AK029057.1 | - | 7 - 8 | 886 | 63 | ||
AK049210.1 | - | 7 - 8 | 916 | 63 | ||
BC120829.1 | - | 7 - 8 | 846 | 40129752 | 63 | |
BC120831.1 | - | 7 - 8 | 846 | 40129754 | 63 | |
BC144835.1 | - | 7 - 8 | 846 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |