Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177152.5 | NP_796126.4 | 13 - 14 | 2060 | 82 | |
GenBank mRNA | AK018522.1 | - | 7 - 8 | 873 | 82 | |
AK129481.1 | - | 9 - 10 | 1009 | 82 | ||
AY505341.1 | - | 13 - 14 | 2003 | 82 | ||
BC065142.1 | - | 13 - 14 | 2026 | 30610561 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |