Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081143.1 | NP_001074612.1 | 2 - 3 | 275 | 81 | |
GenBank mRNA | AK040065.1 | - | 2 - 3 | 276 | 81 | |
BC147449.1 | - | 2 - 3 | 296 | 9056091 | 81 | |
BC147466.1 | - | 2 - 3 | 296 | 9056108 | 81 | |
BC147839.1 | - | 2 - 3 | 224 | 9087556 | 81 | |
BC147841.1 | - | 2 - 3 | 224 | 9087558 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |