Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205036.1 | NP_001191965.1 | 2 - 3 | 288 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons |