Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289657.1 | NP_001276586.1 | 5 - 6 | 1108 | 81 | |
NM_178630.4 | NP_848745.2 | 5 - 6 | 1108 | 81 | ||
GenBank mRNA | AK029798.1 | - | 5 - 6 | 692 | 81 | |
AK030378.1 | - | 5 - 6 | 666 | 81 | ||
AK047649.1 | - | 5 - 6 | 665 | 81 | ||
AK054066.1 | - | 4 - 5 | 2139 | 81 | ||
BC139065.1 | - | 5 - 6 | 560 | 8862087 | 81 | |
DQ867031.1 | - | 5 - 6 | 1109 | 81 | ||
DQ867032.1 | - | 5 - 6 | 1109 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |