Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081412.2 | NP_001074881.1 | 13 - 14 | 2835 | 66 | |
GenBank mRNA | AK129482.1 | - | 10 - 11 | 1025 | 66 | |
AK183908.1 | - | 3 - 4 | 206 | 66 | ||
BC048842.1 | - | 1 - 2 | 79 | 66 | ||
BC060270.1 | - | 13 - 14 | 2073 | 66 | ||
BC131682.1 | - | 14 - 15 | 2240 | 40111128 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |