Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182927.3 | NP_891557.3 | 2 - 3 | 309 | 128 | |
GenBank mRNA | AB115762.1 | - | 1 - 2 | 177 | 128 | |
BC052757.1 | - | 2 - 3 | 281 | 128 | ||
BC062119.1 | - | 2 - 3 | 309 | 6821036 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |