Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009840.3 | NP_033970.3 | 1 - 2 | 138 | 86 | |
GenBank mRNA | AK075880.1 | - | 1 - 2 | 127 | 86 | |
AK145661.1 | - | 1 - 2 | 122 | 86 | ||
AK145754.1 | - | 1 - 2 | 121 | 86 | ||
AK145918.1 | - | 1 - 2 | 140 | 86 | ||
AK147038.1 | - | 1 - 2 | 141 | 86 | ||
AK159969.1 | - | 1 - 2 | 130 | 86 | ||
AK168465.1 | - | 1 - 2 | 110 | 86 | ||
AK172867.1 | - | 1 - 2 | 109 | 86 | ||
BC009007.1 | - | 1 - 2 | 96 | 3582639 | 86 | |
Z37164.1 | - | 1 - 2 | 142 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |