Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000016220.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016220.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016220.mm.1 |
TC1000002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000016221.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016221.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016221.mm.1 |
TC1000002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000016218.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016218.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016218.mm.1 |
TC1000002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000016215.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016215.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016215.mm.1 |
TC1000002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000016216.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016216.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016216.mm.1 |
TC1000002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000016217.mm.1 |
NM_001033260.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016217.mm.1 |
NM_001033260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000016217.mm.1 |
TC1000002266.mm.1 |
MTA TC |