Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213614.2 | NP_998779.2 | 3 - 4 | 240 | 76 | |
GenBank mRNA | AF033350.1 | - | 1 - 2 | 87 | 76 | |
AK134700.1 | - | 3 - 4 | 177 | 76 | ||
BC059848.1 | - | 1 - 2 | 118 | 76 | ||
BC094347.1 | - | 3 - 4 | 213 | 76 | ||
BC141073.1 | - | 3 - 4 | 250 | 9055685 | 76 | |
BC145331.1 | - | 3 - 4 | 250 | 9053887 | 76 | |
BC145332.1 | - | 3 - 4 | 263 | 9053888 | 76 | |
CA945297.1 | - | 3 - 4 | 256 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |