Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164150.1 | NP_001157622.1 | 17 - 18 | 1974 | 65 | |
NM_001164151.1 | NP_001157623.1 | 17 - 18 | 1971 | 65 | ||
NM_001164152.1 | NP_001157624.1 | 17 - 18 | 1971 | 65 | ||
NM_011548.4 | NP_035678.3 | 17 - 18 | 1968 | 65 | ||
GenBank mRNA | AF352579.1 | - | 17 - 18 | 1663 | 65 | |
AK038738.1 | - | 17 - 18 | 1958 | 65 | ||
AK147735.1 | - | 16 - 17 | 1631 | 65 | ||
AK161009.1 | - | 17 - 18 | 1960 | 65 | ||
AK170674.1 | - | 17 - 18 | 1966 | 65 | ||
BC006860.1 | - | 17 - 18 | 1922 | 2631291 | 65 | |
D16631.1 | - | 3 - 4 | 423 | 65 | ||
X17500.1 | - | 4 - 5 | 476 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |