Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174155.1 | NP_001167626.1 | 10 - 11 | 1450 | 85 | |
NM_145149.4 | NP_660131.2 | 10 - 11 | 1450 | 85 | ||
NR_045676.1 | 10 - 11 | 1450 | 85 | |||
GenBank mRNA | AF331457.1 | - | 10 - 11 | 1450 | 85 | |
AK049562.1 | - | 9 - 10 | 1193 | 85 | ||
AK087348.1 | - | 10 - 11 | 1404 | 85 | ||
AK089282.1 | - | 10 - 11 | 1397 | 85 | ||
AK154746.1 | - | 10 - 11 | 1440 | 85 | ||
AK162667.1 | - | 10 - 11 | 1451 | 85 | ||
AK165106.1 | - | 9 - 10 | 1188 | 85 | ||
AY040628.1 | - | 10 - 11 | 1450 | 85 | ||
AY196476.1 | - | 10 - 11 | 1310 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |