Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026815.2 | NP_081091.1 | 5 - 6 | 494 | 92 | |
GenBank mRNA | AF262335.1 | - | 4 - 5 | 466 | 92 | |
AK004014.1 | - | 5 - 6 | 495 | 92 | ||
BC132047.1 | - | 4 - 5 | 486 | 40130324 | 92 | |
BC138390.1 | - | 4 - 5 | 486 | 8861410 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |