Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081429.1 | NP_001074898.1 | 3 - 4 | 695 | 152 | |
GenBank mRNA | AK035850.1 | - | 3 - 4 | 398 | 152 | |
AK041803.1 | - | 3 - 4 | 454 | 152 | ||
AK045055.1 | - | 3 - 4 | 684 | 152 | ||
AK051119.1 | - | 3 - 4 | 661 | 152 | ||
AK133297.1 | - | 3 - 4 | 661 | 152 | ||
AK138231.1 | - | 3 - 4 | 687 | 152 | ||
BC032924.1 | - | 3 - 4 | 457 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |