Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008959.3 | NP_032985.2 | 8 - 9 | 1682 | 63 | |
GenBank mRNA | AF042731.1 | - | 8 - 9 | 1097 | 63 | |
AK036990.1 | - | 8 - 9 | 1144 | 63 | ||
AK054101.1 | - | 8 - 9 | 1121 | 63 | ||
AK085857.1 | - | 8 - 9 | 1683 | 63 | ||
AK086136.1 | - | 8 - 9 | 1341 | 63 | ||
AK088048.1 | - | 8 - 9 | 1164 | 63 | ||
AK137681.1 | - | 8 - 9 | 1352 | 63 | ||
AK172873.1 | - | 7 - 8 | 3468 | 63 | ||
BC003260.1 | - | 8 - 9 | 1319 | 3501141 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |