Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008786.2 | NP_032812.2 | 2 - 3 | 610 | 116 | |
GenBank mRNA | AK006162.1 | - | 2 - 3 | 288 | 116 | |
AK075632.1 | - | 1 - 2 | 97 | 116 | ||
BC040750.1 | - | 2 - 3 | 203 | 116 | ||
BC049613.1 | - | 2 - 3 | 314 | 116 | ||
BC058966.1 | - | 2 - 3 | 299 | 5717963 | 116 | |
D38023.1 | - | 2 - 3 | 274 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |