Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029912.2 | NP_001025083.1 | 11 - 12 | 2518 | 67 | |
GenBank mRNA | AK122519.1 | - | 11 - 12 | 2518 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |