Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011920.3 | NP_036050.1 | 2 - 3 | 652 | 142 | |
GenBank mRNA | AA008579.1 | - | 278 | 142 | ||
AF140218.1 | - | 1 - 2 | 215 | 142 | ||
AK142528.1 | - | 2 - 3 | 298 | 142 | ||
BC053730.1 | - | 2 - 3 | 664 | 30064535 | 142 | |
CO039662.1 | - | 2 - 3 | 142 | |||
KF662864.1 | - | 2 - 3 | 249 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |