Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256432.1 | NP_001243361.1 | 15 - 16 | 2845 | 99 | |
GenBank mRNA | AK134673.1 | - | 11 - 12 | 1473 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase 3' Most Detect Genomic DNA Mus musculus Probe spans exons |