Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007657.3 | NP_031683.1 | 3 - 4 | 386 | 65 | |
GenBank mRNA | AK002251.1 | - | 3 - 4 | 392 | 65 | |
AK012793.1 | - | 3 - 4 | 392 | 65 | ||
AK151443.1 | - | 3 - 4 | 352 | 65 | ||
AK151619.1 | - | 3 - 4 | 339 | 65 | ||
AK170310.1 | - | 3 - 4 | 339 | 65 | ||
BC070474.1 | - | 3 - 4 | 386 | 30634238 | 65 | |
L08115.1 | - | 3 - 4 | 324 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |