Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153589.2 | NP_705817.2 | 1 - 2 | 326 | 150 | |
GenBank mRNA | AK044763.1 | - | 1 - 2 | 327 | 150 | |
FJ384096.1 | - | 1 - 2 | 327 | 150 | ||
KC164759.2 | - | 1 - 2 | 326 | 150 | ||
KC164760.2 | - | 1 - 2 | 326 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |