Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 3 - 4 | 1540 | 67 | |
NM_007992.2 | NP_032018.2 | 3 - 4 | 1540 | 67 | ||
GenBank mRNA | AK159361.1 | - | 3 - 4 | 1852 | 67 | |
AK161522.1 | - | 3 - 4 | 1483 | 67 | ||
AK168688.1 | - | 3 - 4 | 1478 | 67 | ||
BC005443.1 | - | 3 - 4 | 1520 | 3490759 | 67 | |
X75285.1 | - | 3 - 4 | 1440 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |