Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tamm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026894.1 | NP_081170.1 | 2 - 3 | 408 | 82 | |
GenBank mRNA | AK005100.1 | - | 2 - 3 | 408 | 82 | |
AK160774.1 | - | 2 - 3 | 399 | 82 | ||
BB857425.1 | - | 2 - 3 | 410 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |