Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113564.1 | NP_001107036.1 | 4 - 5 | 793 | 126 | |
NM_025814.2 | NP_080090.2 | 4 - 5 | 811 | 126 | ||
GenBank mRNA | AK004678.1 | - | 4 - 5 | 811 | 126 | |
AK011051.1 | - | 4 - 5 | 780 | 126 | ||
AK144764.1 | - | 4 - 5 | 791 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |