Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025467.1 | NP_079743.1 | 2 - 3 | 93 | 99 | |
GenBank mRNA | AK007451.1 | - | 2 - 3 | 93 | 99 | |
AK007705.1 | - | 2 - 3 | 93 | 99 | ||
AK008986.1 | - | 2 - 3 | 94 | 99 | ||
AY943909.1 | - | 2 - 3 | 101 | 99 | ||
BC099445.1 | - | 2 - 3 | 82 | 30788014 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |