Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054041.2 | NP_473382.1 | 8 - 9 | 960 | 65 | |
GenBank mRNA | AF378762.1 | - | 8 - 9 | 900 | 65 | |
AK013005.1 | - | 6 - 7 | 367 | 65 | ||
AK031465.1 | - | 8 - 9 | 950 | 65 | ||
BC094544.1 | - | 8 - 9 | 905 | 6406827 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |