Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053204.2 | NP_444434.2 | 5 - 6 | 1729 | 67 | |
NM_178085.3 | NP_835186.1 | 4 - 5 | 1000 | 67 | ||
NR_104484.1 | 5 - 6 | 1729 | 67 | |||
GenBank mRNA | AF340028.1 | - | 5 - 6 | 1713 | 67 | |
AF340029.1 | - | 5 - 6 | 1713 | 67 | ||
AK035989.1 | - | 4 - 5 | 1543 | 67 | ||
AK048990.1 | - | 4 - 5 | 953 | 67 | ||
AY316692.1 | - | 3 - 4 | 1161 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |