Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001179.3 | NP_001001179.2 | 14 - 15 | 1762 | 73 | |
GenBank mRNA | AK138836.1 | - | 14 - 15 | 1763 | 73 | |
AK161068.1 | - | 1 - 2 | 112 | 73 | ||
AK161184.1 | - | 14 - 15 | 1765 | 73 | ||
BC040781.1 | - | 2 - 3 | 214 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |