Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 16 - 17 | 2948 | 84 | |
GenBank mRNA | AK129082.1 | - | 16 - 17 | 2908 | 84 | |
AK133013.1 | - | 16 - 17 | 2898 | 84 | ||
AK162178.1 | - | 16 - 17 | 3006 | 84 | ||
BC030906.1 | - | 9 - 10 | 1748 | 84 | ||
BC057054.1 | - | 16 - 17 | 2948 | 6827225 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |