Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133345.2 | NP_579923.1 | 3 - 4 | 293 | 79 | |
GenBank mRNA | AK002821.1 | - | 3 - 4 | 295 | 79 | |
AK009267.1 | - | 3 - 4 | 294 | 79 | ||
AK031633.1 | - | 3 - 4 | 298 | 79 | ||
AK050522.1 | - | 3 - 4 | 306 | 79 | ||
AY035880.1 | - | 3 - 4 | 280 | 79 | ||
AY035881.1 | - | 3 - 4 | 280 | 79 | ||
AY036107.1 | - | 3 - 4 | 280 | 79 | ||
BC009127.1 | - | 3 - 4 | 281 | 3708722 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |