Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029549.mm.1 |
TC0400003611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029549.mm.1 |
NM_199305.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029549.mm.1 |
NM_199305.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029553.mm.1 |
TC0400003611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029553.mm.1 |
NM_199305.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029553.mm.1 |
NM_199305.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029554.mm.1 |
TC0400003611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029554.mm.1 |
NM_199305.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029554.mm.1 |
NM_199305.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029550.mm.1 |
TC0400003611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029550.mm.1 |
NM_199305.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029550.mm.1 |
NM_199305.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029552.mm.1 |
TC0400003611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029552.mm.1 |
NM_199305.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029552.mm.1 |
NM_199305.1 |
Refseq |