Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159581.1 | NP_001153053.1 | 10 - 11 | 840 | 90 | |
NM_133250.2 | NP_573513.2 | 9 - 10 | 747 | 90 | ||
GenBank mRNA | AB117937.1 | - | 7 - 8 | 635 | 90 | |
AB117938.1 | - | 8 - 9 | 709 | 90 | ||
AK049542.1 | - | 1 - 2 | 89 | 90 | ||
AK077546.1 | - | 9 - 10 | 744 | 90 | ||
AK156790.1 | - | 10 - 11 | 830 | 90 | ||
AY007717.1 | - | 8 - 9 | 703 | 90 | ||
BC057942.1 | - | 9 - 10 | 728 | 5320744 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |