Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243043.1 | NP_001229972.1 | 17 - 18 | 2770 | 129 | |
NM_001243044.1 | NP_001229973.1 | 16 - 17 | 2552 | 129 | ||
NM_007454.3 | NP_031480.2 | 16 - 17 | 2552 | 129 | ||
GenBank mRNA | AA003413.1 | - | 2 - 3 | 188 | 129 | |
AK034140.1 | - | 17 - 18 | 2773 | 129 | ||
AK155892.1 | - | 16 - 17 | 2472 | 129 | ||
AK159273.1 | - | 16 - 17 | 2443 | 129 | ||
AK160042.1 | - | 16 - 17 | 2443 | 129 | ||
AK165747.1 | - | 16 - 17 | 2470 | 130 | ||
AK170376.1 | - | 16 - 17 | 2463 | 129 | ||
BC008513.1 | - | 16 - 17 | 2365 | 3601435 | 129 | |
BC066827.1 | - | 7 - 8 | 1096 | 129 | ||
Y07919.1 | - | 16 - 17 | 2407 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |