Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009557.3 | NP_033583.2 | 1 - 2 | 414 | 57 | |
GenBank mRNA | AK051031.1 | - | 1 - 2 | 387 | 57 | |
AK053652.1 | - | 1 - 2 | 417 | 57 | ||
AK136405.1 | - | 1 - 2 | 323 | 57 | ||
AK148255.1 | - | 1 - 2 | 363 | 57 | ||
AK173240.1 | - | 1 - 2 | 401 | 57 | ||
BB665308.1 | - | 1 - 2 | 414 | 57 | ||
BC006587.1 | - | 1 - 2 | 81 | 3587306 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |