Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020000.3 | NP_064384.2 | 2 - 3 | 167 | 113 | |
GenBank mRNA | AB041805.1 | - | 1 - 2 | 1623 | 113 | |
AK008759.1 | - | 2 - 3 | 146 | 113 | ||
AK147032.1 | - | 2 - 3 | 153 | 113 | ||
AK148025.1 | - | 2 - 3 | 148 | 113 | ||
BC002315.1 | - | 1 - 2 | 107 | 113 | ||
BC021870.1 | - | 2 - 3 | 138 | 4022630 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |