Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
TC0600003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_001204239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_001204239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_001204253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_001204253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_019985.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012484.mm.1 |
NM_019985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
TC0600003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_001204239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_001204239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_001204253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_001204253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_019985.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012488.mm.1 |
NM_019985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
TC0600003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_001204239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_001204239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_001204253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_001204253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_019985.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600012486.mm.1 |
NM_019985.3 |
Refseq |