Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177758.4 | NP_808426.2 | 1 - 2 | 102 | 101 | |
GenBank mRNA | AK144249.1 | - | 1 - 2 | 103 | 101 | |
AK153688.1 | - | 1 - 2 | 39 | 101 | ||
AK153845.1 | - | 1 - 2 | 39 | 101 | ||
AK162787.1 | - | 1 - 2 | 39 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |