Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 4 - 5 | 1266 | 102 | |
NM_001025393.1 | NP_001020564.1 | 4 - 5 | 1266 | 102 | ||
NM_153787.2 | NP_722482.1 | 4 - 5 | 1260 | 102 | ||
GenBank mRNA | AK088741.1 | - | 4 - 5 | 1201 | 102 | |
AK129071.1 | - | 1 - 2 | 550 | 102 | ||
AK141834.1 | - | 4 - 5 | 1213 | 102 | ||
AK149724.1 | - | 4 - 5 | 1219 | 102 | ||
AK150017.1 | - | 4 - 5 | 1219 | 102 | ||
AK161608.1 | - | 156 | 102 | |||
AK161692.1 | - | 4 - 5 | 1264 | 102 | ||
AK162896.1 | - | 4 - 5 | 1224 | 102 | ||
BC028963.1 | - | 4 - 5 | 1198 | 102 | ||
BC029682.1 | - | 4 - 5 | 1197 | 102 | ||
BC034300.1 | - | 4 - 5 | 1192 | 4507876 | 102 | |
BC086624.1 | - | 4 - 5 | 1248 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |