Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172439.3 | NP_766027.2 | 11 - 12 | 2471 | 64 | |
GenBank mRNA | AK034272.1 | - | 11 - 12 | 2472 | 64 | |
AK136908.1 | - | 11 - 12 | 2458 | 64 | ||
AK170114.1 | - | 11 - 12 | 2437 | 64 | ||
AK171029.1 | - | 11 - 12 | 2432 | 64 | ||
BC022915.1 | - | 11 - 12 | 2424 | 64 | ||
BC046561.1 | - | 10 - 11 | 1284 | 64 | ||
BC051452.1 | - | 11 - 12 | 2440 | 64 | ||
BC131634.1 | - | 11 - 12 | 2466 | 40096940 | 64 | |
BC131635.1 | - | 11 - 12 | 2463 | 40096943 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |