Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161845.2 | NP_001155317.2 | 1 - 2 | 526 | 80 | |
GenBank mRNA | BC070401.1 | - | 1 - 2 | 526 | 5695396 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |